Thermo Scientific™ MAS™ Immunology Controls, Thermo Scientific
Supplier: ISE Ilmenauer Systemeinrichtungen
IC-202 IC-303 IC-101
89130-636EA 1111.11 USD InStock
89130-636 89130-638 89130-634
Thermo Scientific™ MAS™ Immunology Controls, Thermo Scientific
Controls and Calibration Verification Materials
Monitor the performance of various serum protein and serology test procedures with ready-to-use, liquid assayed multi-analyte MAS™ Immunology Controls
Assayed for most major instrument systems. Three levels provide monitoring of the full assay range. Contains specific serum proteins, including Antistreptolysin, CRP, Prealbumin, and Rheumatoid Factor. Human-serum matrix.
Analytes: Albumin, a-1-Acid Glycoprotein, a-1-Antitrypsin, a-2-Macroglobulin, Antistreptolysin, Antithrombin III, APO A (HDL Concentrate), APO B (LDL Concentrate), B-2-microglobulin, C3 Complement, C4 Complement, Ceruloplasmin, Haptoglobin, IgA, IgE, IgG, IgM, Kappa Light Chain, Lambda Light Chain, Prealbumin, Properdin B, Rheumatoid Factor, Total Protein, Transferrin. (refer to package insert for specific analyte and stability claims).
These controls are also available in a convenient Vista™ vial format.
Información para pedidos: Available in individual levels only.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty