Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trimethylsilylacetylene ≥97%

Danger

14-8200-25G 14-8200-5G
100208-800EA 349.6 USD InStock
100208-800 100208-802
Trimethylsilylacetylene ≥97%
Trimethylsilylacetylene


Formula: C₅H₁₀Si
Boiling Pt: 53 °C
Density: 0.7111 g/cm³ (20 °C)
Flash Pt: <–30 °F
MDL Number: MFCD00008569
CAS Number: 1066-54-2
EINECS: 213-919-9
UN: 1993
ADR: 3,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty