Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dicyclohexylphosphine ≥98%

Danger

15-1120-50G 15-1120-10G
100200-424EA 796.86 USD InStock
100200-424 100200-422
Dicyclohexylphosphine ≥98%
Dicyclohexylphosphine
Phosphine


Formula: C₁₂H₂₃P
Boiling Pt: 281 - 283 °C (1013 hPa)
Density: 0.904 g/cm³ (20 °C)
MDL Number: MFCD00014296
CAS Number: 829-84-5
EINECS: 212-590-9
UN: 2845
ADR: 4.2,I
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty