Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tri-tert-butylphosphine ≥99%

Danger

15-5810-1G 15-5810-5G
100198-132EA 96.25 USD InStock
100198-132 100198-134
Tri-tert-butylphosphine ≥99%
Tri-tert-butylphosphine
Phosphine


Formula: C₁₂H₂₇P
MW: 202.32 g/mol
Boiling Pt: 102…103 °C (13 mmHg)
Melting Pt: 30 °C
Density: 0.812
Flash Pt: 1 °F
MDL Number: MFCD00015006
CAS Number: 13716-12-6
EINECS: 237-266-4
UN: 2845
ADR: 4.2 I
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty