Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Triisopropylphosphine 10% (w/w) in hexanes ≥98%

Danger

15-6954-50G 15-6954-10G 15-6954-250G
100198-146EA 440.18 USD InStock
100198-146 100204-984 100198-144
Triisopropylphosphine 10% (w/w) in hexanes ≥98%
Triisopropylphosphine
Phosphine


Formula: C₉H₂₁P
MDL Number: MFCD00015029
CAS Number: 6476-36-4
EINECS: 229-333-1
UN: 1993
ADR: 4.2,I
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty