Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tris(2,4,6-trimethoxyphenyl)phosphine ≥97%

Warning

15-8015-10G 15-8015-2G
100199-362EA 368 USD InStock
100199-362 100204-986
Tris(2,4,6-trimethoxyphenyl)phosphine ≥97%
Tris(2,4,6-trimethoxyphenyl)phosphine


Formula: C₂₇H₃₃O₉P
Melting Pt: 155…160 °C
MDL Number: MFCD00145467
CAS Number: 91608-15-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty