Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tris(4-methylphenyl)phosphine ≥98%

Warning

15-8120-25G 15-8120-5G
100200-876EA 345.35 USD InStock
100200-876 100207-604
Tris(4-methylphenyl)phosphine ≥98%
Tris(4-methylphenyl)phosphine
Phosphine


Formula: C₂₁H₂₁P
MW: 304.37 g/mol
Boiling Pt: 400
Melting Pt: 145…148 °C
Density: 1.11 g/cm³ (20 °C)
MDL Number: MFCD00008542
CAS Number: 1038-95-5
EINECS: 213-863-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty