Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ammonium perrhenate ≥99.999% (Re basis), PURATREM

Danger

02-0900-1G 02-0900-5G
100200-366EA 87.75 USD InStock
100200-366 100202-344
Ammonium perrhenate ≥99.999% (Re basis), PURATREM
Ammonium perrhenate


Formula: H₄NO₄Re
Melting Pt: Dec.
Density: 3.97 g/cm³ (20 °C)
MDL Number: MFCD00012377
CAS Number: 13598-65-7
EINECS: 237-075-6
UN: 1479
ADR: 5.1,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty