Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nickel(II) acetylacetonate hydrate


28-1110-500G 28-1110-100G
100210-806EA 363.75 USD InStock
100210-806 100201-570
Nickel(II) acetylacetonate hydrate
Nickel(II) acetylacetonate hydrate
Metal Beta-diketonates, Volatile Precursors for CVD


Formula: C₁₀H₁₄NiO₄·xH₂O
MW: 256.91 g/mol (anhy)
MDL Number: MFCD00149058
CAS Number: 120156-44-7
EINECS: 221-875-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty