Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Zinc bromide ≥99.999% (Zn basis), PURATREM

Danger

30-3033-25G 30-3033-5G
100205-838EA 182.58 USD InStock
100205-838 100203-088
Zinc bromide ≥99.999% (Zn basis), PURATREM
Zinc bromide


Formula: Br₂Zn
MW: 225.2 g/mol
Boiling Pt: 650 °C
Melting Pt: 394
Density: 4.201
Storage Temperature: Ambient
MDL Number: MFCD00011294
CAS Number: 7699-45-8
EINECS: 231-718-4
UN: 1759
ADR: 8 II
Merck Index: 13,10182
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty