Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lithium hexafluoroarsenate ≥99.9% (As basis)

Danger

03-1250-25G 03-1250-5G
100195-902EA 441.6 USD InStock
100195-902 100206-182
Lithium hexafluoroarsenate ≥99.9% (As basis)
Lithium hexafluoroarsenate


Formula: AsF₆Li
MW: 195.85
Melting Pt: 349
MDL Number: MFCD00011076
CAS Number: 29935-35-1
EINECS: 249-963-0
UN: 1557
ADR: 6.1 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty