Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Zirconium ≥99.8%, foil


40-0070-50X50MM 40-0070-100X425MM 40-0070-100X100MM
100204-046EA 106.15 USD InStock
100204-046 100196-944 100200-936
Zirconium ≥99.8%, foil
Zirconium


Formula: Zr
MW: 91.22 g/mol
Boiling Pt: 3577 °C
Melting Pt: 1852 °C
Density: 6.506
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty