Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Molybdenum pentachloride, anhydrous ≥99.6%

Danger

42-1390-50G 42-1390-250G 42-1390-1000G
100206-620EA 300.06 USD InStock
100206-620 100206-618 101324-694
Molybdenum pentachloride, anhydrous ≥99.6%
Molybdenum pentachloride


Formula: Cl₅Mo
Boiling Pt: 268 °C
Melting Pt: 194 °C
Density: 2.928 g/cm³ (20 °C)
MDL Number: MFCD00003467
CAS Number: 10241-05-1
EINECS: 233-575-3
UN: 2508
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty