Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrapropylammonium perruthenate ≥97%

Danger

44-7850-500MG 44-7850-2G
100209-952EA 158.52 USD InStock
100209-952 100198-794
Tetrapropylammonium perruthenate ≥97%
Tetrapropylammonium perruthenate


Formula: C₁₂H₂₈NO₄Ru
Melting Pt: 165 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00074914
CAS Number: 114615-82-6
UN: 1479
ADR: 5.1,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty