Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Indium(III) fluoride, anhydrous ≥98%

Warning

49-1500-5G 49-1500-25G
100208-930EA 188.25 USD InStock
100208-930 100203-798
Indium(III) fluoride, anhydrous ≥98%
Indium(III) fluoride


Formula: F₃In
Boiling Pt: > 1200 °C
Melting Pt: ~1170 °C
MDL Number: MFCD00016149
CAS Number: 7783-52-0
EINECS: 232-005-0
UN: 2923
ADR: 8,II
Merck Index: 12,04990
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty