Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Tolylboronic acid ≥97%

Warning

05-1050-25G 05-1050-5G
100208-254EA 690.71 USD InStock
100208-254 100200-374
2-Tolylboronic acid ≥97%
2-Tolylboronic acid
Boronic Acid and Ester


Formula: C₇H₉BO₂
MW: 135.96 g/mol
Melting Pt: 162…164 °C
MDL Number: MFCD00093526
CAS Number: 16419-60-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty