Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Barium ≥99% (Sr <1.6%), rods

Danger

56-0080-500G 56-0080-100G
100207-412EA 1268.18 USD InStock
100207-412 100201-006
Barium ≥99% (Sr <1.6%), rods
Barium


Formula: Ba
MW: 137.33
Boiling Pt: 1640
Melting Pt: 725
Density: 3.51
Storage Temperature: Ambient
MDL Number: MFCD00134031
CAS Number: 7440-39-3
EINECS: 231-149-1
UN: 1400
ADR: 4.3 II
Merck Index: 12,00991
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty