Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Barium nitrate ≥99.999% (Ba basis), PURATREM

Warning

56-7500-10G 56-7500-50G
100208-452EA 62.28 USD InStock
100208-452 100198-224
Barium nitrate ≥99.999% (Ba basis), PURATREM
Barium nitrate


Formula: Ba(NO₃)₂
Melting Pt: 592 °C
Density: ~ 3.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003442
CAS Number: 10022-31-8
EINECS: 233-020-5
UN: 1446
ADR: 5.1,II
Merck Index: 13,00986
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty