Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cerium tetrafluoride hydrate

Warning

58-0505-50G 58-0505-10G
100201-710EA 397.72 USD InStock
100201-710 100204-092
Cerium tetrafluoride hydrate
Cerium tetrafluoride hydrate


Formula: CeF₄·H₂O
MW: 234.12
Melting Pt: 650 °C (dec.)
MDL Number: MFCD00049422
CAS Number: 60627-09-0
UN: 3288
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty