Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ytterbium(III) fluoride 99,99 Yb basis, PURATREM

Warning

70-7004-5G 70-7004-25G
100199-434EA 161.35 USD InStock
100199-434 100206-786
Ytterbium(III) fluoride 99,99 Yb basis, PURATREM
Ytterbium(III) fluoride


Formula: F₃Yb
MW: 230.04
Boiling Pt: 2227
Melting Pt: 1157 °C
Density: 8.168
MDL Number: MFCD00049615
CAS Number: 13760-80-0
EINECS: 237-354-2
UN: 3288
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty