Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Osmium tetraoxide ≥99.95% (Os basis)

Danger

76-2950-5G 76-2950-500MG 76-2950-1G 76-2950-250MG
100204-648EA 833.66 USD InStock
100204-648 100199-448 100202-688 100201-736
Osmium tetraoxide ≥99.95% (Os basis)
Osmium tetraoxide


Formula: O₄Os
MW: 254.23 g/mol
Boiling Pt: 130
Melting Pt: 40
Density: 4.906
Storage Temperature: Ambient
MDL Number: MFCD00011150
CAS Number: 20816-12-0
EINECS: 244-058-7
UN: 2471
ADR: 6.1 I
Merck Index: 13,06962
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty