Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fluorobenzene ≥99%

Danger

09-3480-100G 09-3480-25G 09-3480-500G
100207-538EA 45.29 USD InStock
100207-538 100209-592 100208-772
Fluorobenzene ≥99%
Fluorobenzene
Organo Fluorine


Formula: C₆H₅F
Boiling Pt: 84…85 °C
Melting Pt: –42 °C
Density: 1.023 g/cm³ (20 °C)
Flash Pt: 9 °F
Storage Temperature: Ambient
MDL Number: MFCD00000280
CAS Number: 462-06-6
EINECS: 207-321-7
UN: 2387
ADR: 3,II
Merck Index: 13,04197
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty