Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6,6,7,7,8,8,8-Heptafluoro-2,2-dimethyl-3,5-octanedione ≥98%

Warning

09-4496-5G 09-4496-25G
100207-966EA 141.54 USD InStock
100207-966 100207-964
6,6,7,7,8,8,8-Heptafluoro-2,2-dimethyl-3,5-octanedione ≥98%
6,6,7,7,8,8,8-Heptafluoro-2,2-dimethyl-3,5-octanedione
Organo Fluorine


Formula: C₁₀H₁₁F₇O₂
Boiling Pt: 56…57 °C (10 mmHg)
Melting Pt: 34…35 °C
Density: 1.27 g/cm³ (20 °C)
Flash Pt: 101 °F
Storage Temperature: Ambient
MDL Number: MFCD00000435
CAS Number: 17587-22-3
EINECS: 241-556-6
UN: 1993
ADR: 3,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty