Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexafluoroglutaric acid ≥97%

Danger

09-4700-5G 09-4700-25G
100203-926EA 107.57 USD InStock
100203-926 100210-968
Hexafluoroglutaric acid ≥97%
Hexafluoroglutaric acid
Organo Fluorine


Formula: C₅H₂F₆O₄
Boiling Pt: 134…138 °C (3 mmHg)
Melting Pt: 92…97 °C
MDL Number: MFCD00004172
CAS Number: 376-73-8
EINECS: 206-813-9
UN: 3261
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty