Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ammonium chloride ≥99.999%, PURATREM

Warning

93-0242-100G 93-0242-25G
100208-988EA 263.26 USD InStock
100208-988 100205-226
Ammonium chloride ≥99.999%, PURATREM
Ammonium chloride


Formula: NH₄Cl
Boiling Pt: 520 °C
Melting Pt: 340 °C (subl.)
Density: 1.5256 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011420
CAS Number: 12125-02-9
EINECS: 235-186-4
Merck Index: 13,00510
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e45c4909ae60875d5b9dca2f28f54f5c8028e64&vid=28e617a1f562ee2a112e01765ea798627493a5f1&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty