Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lithium amide ≥95%

Danger

93-0306-250G 93-0306-50G
100208-070EA 315.63 USD InStock
100208-070 100205-230
Lithium amide ≥95%
Lithium amide


Formula: H₂LiN
Boiling Pt: 430 °C (1013 hPa)
Melting Pt: 373 °C
Density: 1.17 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011093
CAS Number: 7782-89-0
EINECS: 231-968-4
UN: 1390
ADR: 4.3,II
Merck Index: 13,05544
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty