Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tributyl borate ≥98%

Warning

93-0528-500G 93-0528-100G
100208-490EA 230.71 USD InStock
100208-490 100205-260
Tributyl borate ≥98%
Tributyl borate


Formula: C₁₂H₂₇BO₃
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: -70 °C
Density: 0.8567 g/cm³ (20 °C)
Flash Pt: 200 °F
MDL Number: MFCD00009434
CAS Number: 688-74-4
EINECS: 211-706-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty