Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trimethyl borate 98

Danger

93-0531-500G 93-0531-2000G
100197-642EA 72.18 USD InStock
100197-642 100210-132
Trimethyl borate 98
Trimethyl borate


Formula: C₃H₉BO₃
MW: 103.91 g/mol
Boiling Pt: 67
Melting Pt: -5
Density: 0.915
Flash Pt: 30 °F
Storage Temperature: Refrigerator
MDL Number: MFCD00008346
CAS Number: 121-43-7
EINECS: 204-468-9
UN: 2416
ADR: 3 II
Merck Index: 12,09842
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty