Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium tetraphenylborate ≥99.5% ACS

Danger

93-0571-100G 93-0571-5G 93-0571-25G
100200-030EA 450.09 USD InStock
100200-030 100201-826 100201-824
Sodium tetraphenylborate ≥99.5% ACS
Sodium tetraphenylborate


Formula: (C₆H₅)₄BNa
MW: 342.22 g/mol
Melting Pt: > 400 °C
Density: 1.15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011494
CAS Number: 143-66-8
EINECS: 205-605-5
UN: 2811
ADR: 6.1 III
Merck Index: 13,08765
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty