Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium hexafluoroarsenate ≥99%

Danger

93-1145-25G 93-1145-5G
100200-038EA 410.46 USD InStock
100200-038 100206-904
Sodium hexafluoroarsenate ≥99%
Sodium hexafluoroarsenate


Formula: AsF₆Na
MDL Number: MFCD00014228
CAS Number: 12005-86-6
UN: 1557
ADR: 6.1,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty