Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

penta-Sodium triphosphate ≥85%, Technical Grade

Warning

93-1195-500G 93-1195-2000G
100205-292EA 48.12 USD InStock
100205-292 100204-668
penta-Sodium triphosphate ≥85%, Technical Grade
penta-Sodium triphosphate


Formula: Na₅O₁₀P₃
MW: 367.86 g/mol
Melting Pt: 622
Density: 2.52 g/cm³ (20 °C)
MDL Number: MFCD00003514
CAS Number: 7758-29-4
EINECS: 231-838-7
Merck Index: 12,08846
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty