Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Magnesium bromide hexahydrate ≥98%

Warning

93-1219-1000G 93-1219-250G
100201-256EA 509.54 USD InStock
100201-256 100207-768
Magnesium bromide hexahydrate ≥98%
Magnesium bromide hexahydrate


Formula: Br₂Mg·6H₂O
Melting Pt: 172 °C
Density: 2 g/cm³ (20 °C)
MDL Number: MFCD00149780
CAS Number: 13446-53-2
EINECS: 232-170-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty