Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Aluminium oxide activated neutral, γ-phase ≥96%

Warning

93-1338-500G 93-1338-2000G
100196-494EA 106.15 USD InStock
100196-494 100199-482
Aluminium oxide activated neutral, γ-phase ≥96%
Aluminium oxide activated neutral, γ-phase


Formula: Al₂O₃
Boiling Pt: 2980 °C
Melting Pt: 2045 °C
Density: 3.97 g/cm³ (20 °C)
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
Merck Index: 12,00369
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty