Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Aluminium iodide ≥95%

Danger

93-1389-50G 93-1389-10G
100210-680EA 317.05 USD InStock
100210-680 100201-904
Aluminium iodide ≥95%
Aluminium iodide


Formula: AlI₃
MW: 407.7
Boiling Pt: 382
Melting Pt: 191
Density: 3.98
Storage Temperature: Ambient
MDL Number: MFCD00003427
CAS Number: 7784-23-8
EINECS: 232-054-8
UN: 3260
ADR: 8 II
Merck Index: 12,00358
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty