Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium hexafluoroaluminate ≥99.9% (Al basis)

Danger

93-1399-10G 93-1399-2G
100199-486EA 161.35 USD InStock
100199-486 100205-324
Sodium hexafluoroaluminate ≥99.9% (Al basis)
Sodium hexafluoroaluminate


Formula: AlF₆Na₃
Melting Pt: 977 °C
Density: 2.95 g/cm³ (20 °C)
MDL Number: MFCD00003507
CAS Number: 13775-53-6
EINECS: 237-410-6
UN: 3260
ADR: 8,III
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty