Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethoxytrimethylsilane ≥97%

Danger

93-1465-100G 93-1465-25G
100205-348EA 319.88 USD InStock
100205-348 100196-510
Ethoxytrimethylsilane ≥97%
Ethoxytrimethylsilane


Formula: C₅H₁₄OSi
MW: 118.25 g/mol
Boiling Pt: 75
Melting Pt: -83
Density: 0.757
Flash Pt: –4 °F
MDL Number: MFCD00009069
CAS Number: 1825-62-3
EINECS: 217-370-6
UN: 1993
ADR: 3 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty