Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyldichlorosilane ≥99%

Danger

93-1493-250G 93-1493-1000G
100210-208EA 86.34 USD InStock
100210-208 100205-862
Dimethyldichlorosilane ≥99%
Dimethyldichlorosilane


Formula: C₂H₆Cl₂Si
MW: 129.06 g/mol
Boiling Pt: 70
Melting Pt: -76
Density: 1.070
Flash Pt: 3 °F
MDL Number: MFCD00000491
CAS Number: 75-78-5
EINECS: 200-901-0
UN: 1162
ADR: 3 II
REACH: 01-2119437250-51
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty