Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexamethyldisilane ≥97%

Danger

93-1494-25G 93-1494-5G
100205-360EA 182.58 USD InStock
100205-360 100208-508
Hexamethyldisilane ≥97%
Hexamethyldisilane


Formula: C₆H₁₈Si₂
MW: 146.38 g/mol
Boiling Pt: 112…113 °C
Melting Pt: 13…15 °C
Density: 0.715
Flash Pt: 29 °F
Storage Temperature: Refrigerator
MDL Number: MFCD00008258
CAS Number: 1450-14-2
EINECS: 215-911-0
UN: 1993
ADR: 3 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty