Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Triethyl phosphate ≥99%

Warning

93-1559-500G 93-1559-2000G
100197-716EA 90.58 USD InStock
100197-716 100211-218
Triethyl phosphate ≥99%
Triethyl phosphate
Volatile Precursors for CVD


Formula: C₆H₁₅O₄P
MW: 182.16 g/mol
Boiling Pt: 215
Melting Pt: -57
Density: 1.072
Flash Pt: 240 °F
Storage Temperature: Ambient
MDL Number: MFCD00009077
CAS Number: 78-40-0
EINECS: 201-114-5
Merck Index: 12,09806
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty