Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Calcium iodide, anhydrous ≥98%

Warning

93-2024-100G 93-2024-25G
100199-506EA 714.77 USD InStock
100199-506 100206-954
Calcium iodide, anhydrous ≥98%
Calcium iodide


Formula: CaI₂
MW: 293.89 g/mol
Boiling Pt: 1100
Melting Pt: 740
Density: 3.956 g/cm³ (25 °C)
MDL Number: MFCD00010910
CAS Number: 10102-68-8
EINECS: 233-276-8
Merck Index: 12,01720
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty