Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Calcium ≥99%, turnings

Danger

93-2077-250G 93-2077-50G
100207-460EA 426.03 USD InStock
100207-460 100197-734
Calcium ≥99%, turnings
Calcium


Formula: Ca
MW: 40.08 g/mol
Boiling Pt: 1484
Melting Pt: 850
Density: 1.54
Storage Temperature: Ambient
MDL Number: MFCD00085314
CAS Number: 7440-70-2
EINECS: 231-179-5
UN: 1401
ADR: 4.3 II
Merck Index: 13,01644
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty