Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium metavanadate ≥96%

Danger

93-2303-500G 93-2303-100G
100202-536EA 608.62 USD InStock
100202-536 100209-048
Sodium metavanadate ≥96%
Sodium metavanadate


Formula: NaO₃V
Melting Pt: 630 °C
Density: 5.15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011125
CAS Number: 13718-26-8
EINECS: 237-272-7
UN: 3288
ADR: 6.1,III
Merck Index: 12,08849
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty