Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Manganese ≥99.5%, powder

Warning

93-2543-1000G 93-2543-250G
100210-286EA 309.97 USD InStock
100210-286 100205-454
Manganese ≥99.5%, powder
Manganese


Formula: Mn
MW: 54.94 g/mol
Boiling Pt: 1900 °C
Melting Pt: 1260 °C
Density: 7.20
MDL Number: MFCD00011111
CAS Number: 7439-96-5
EINECS: 231-105-1
UN: 3089
ADR: 4.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty