Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Copper(I) acetate ≥97%

Warning

93-2989-50G 93-2989-10G
100201-094EA 557.66 USD InStock
100201-094 100201-092
Copper(I) acetate ≥97%
Copper(I) acetate


Formula: C₂H₃CuO₂
MW: 122.59 g/mol
Melting Pt: 250
MDL Number: MFCD00058908
CAS Number: 598-54-9
EINECS: 209-938-7
Merck Index: 12,02728
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty