Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Copper ≥99.5%, powder

Warning

93-2999-2000G 93-2999-100G 93-2999-500G
100209-088EA 338.28 USD InStock
100209-088 100197-812 100200-750
Copper ≥99.5%, powder
Copper


Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.92
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
UN: 3089
ADR: 4.1 II
Merck Index: 13,02545
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty