Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl phosphonate ≥95%

Warning

93-1521-100G 93-1521-500G
101433-276EA 72.18 USD InStock
101433-276 100201-070
Diethyl phosphonate ≥95%
Diethyl phosphonate


Formula: C₄H₁₁O₃P
MW: 138.1 g/mol
Boiling Pt: 50
Density: 1.072
Flash Pt: 195 °F
MDL Number: MFCD00044573
CAS Number: 762-04-9
EINECS: 212-091-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty