Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Zinc selenide ≥99.999% (Zn basis), PURATREM

Danger

93-3023-50G 93-3023-10G
100207-286EA 578.89 USD InStock
100207-286 100200-752
Zinc selenide ≥99.999% (Zn basis), PURATREM
Zinc selenide


Formula: SeZn
Melting Pt: > 1100 °C
Density: 5.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011296
CAS Number: 1315-09-9
EINECS: 215-259-7
UN: 3283
ADR: 6.1,III
Merck Index: 12,10290
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty