Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Zinc(II) acetylacetonate hydrate ≥98%

Warning

93-3031-50G 93-3031-250G
100199-068EA 45.29 USD InStock
100199-068 100202-550
Zinc(II) acetylacetonate hydrate ≥98%
Zinc(II) acetylacetonate hydrate


Formula: C₁₀H₁₄O₄Zn·H₂O
MW: 281.62
MDL Number: MFCD00149060
CAS Number: 108503-47-5
EINECS: 237-860-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty