Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Arsenic(III) chloride ≥99.999% (As basis), PURATREM

Danger

93-3309-25G 93-3309-5G 93-3309-100G
100205-528EA 225.05 USD InStock
100205-528 100202-098 100210-688
Arsenic(III) chloride ≥99.999% (As basis), PURATREM
Arsenic(III) chloride


Formula: AsCl₃
MW: 181.28
Boiling Pt: 130.1
Melting Pt: -8.5
Density: 2.163
MDL Number: MFCD00014165
CAS Number: 7784-34-1
EINECS: 232-059-5
UN: 1560
ADR: 6.1 I
Merck Index: 12,00841
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty