Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Rubidium bromide ≥99.9% (Rb basis)


93-3726-5G 93-3726-25G
100203-456EA 94.83 USD InStock
100203-456 100210-690
Rubidium bromide ≥99.9% (Rb basis)
Rubidium bromide


Formula: BrRb
Boiling Pt: 1340 °C
Melting Pt: 682 °C
Density: 3.35 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011186
CAS Number: 7789-39-1
EINECS: 232-162-5
Merck Index: 12,08440
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty